siRNA Details
Gene Symbol: Tlr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207604.1 | NP_997487.1 | 2 | 1067 | ||
XM_006250960.2 | XP_006251022.1 | 2 | 923 | |||
GenBank mRNA | BC062390.1 | - | 2 | 1067 | 5621142 |
