siRNA Details
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107220.1 | NP_001100690.1 | 25 | 2041 | ||
GenBank mRNA | BC086546.1 | - | 15 | 955 | ||
FQ227240.1 | - | 7 | 473 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107220.1 | NP_001100690.1 | 25 | 2041 | ||
GenBank mRNA | BC086546.1 | - | 15 | 955 | ||
FQ227240.1 | - | 7 | 473 |