siRNA Details
Gene Symbol: Spred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047094.1 | NP_001040559.1 | 657 | |||
GenBank mRNA | AB125136.1 | - | 657 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047094.1 | NP_001040559.1 | 657 | |||
GenBank mRNA | AB125136.1 | - | 657 |