siRNA Details
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127537.1 | NP_001121009.1 | 12 | 1866 | ||
GenBank mRNA | BC161962.1 | - | 12 | 1866 | 9027983 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127537.1 | NP_001121009.1 | 12 | 1866 | ||
GenBank mRNA | BC161962.1 | - | 12 | 1866 | 9027983 |