siRNA Details
Gene Symbol: Socs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107256.2 | NP_001100726.1 | 2 | 272 | ||
GenBank mRNA | BC168879.1 | - | 2 | 272 | 9035196 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Socs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107256.2 | NP_001100726.1 | 2 | 272 | ||
GenBank mRNA | BC168879.1 | - | 2 | 272 | 9035196 |