siRNA Details
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170468.1 | NP_001163939.1 | 18 | 3699 | ||
XM_006251958.2 | XP_006252020.1 | 18 | 3632 | |||
XM_006251959.2 | XP_006252021.1 | 18 | 3629 | |||
XM_006251960.2 | XP_006252022.1 | 17 | 3548 | |||
XM_006251962.2 | XP_006252024.1 | 17 | 3357 | |||
XM_006251963.1 | XP_006252025.1 | 11 | 1672 | |||
XM_006251964.1 | XP_006252026.1 | 11 | 1454 | |||
GenBank mRNA | BC091320.1 | - | 9 | 1192 | ||
CF110347.1 | - | 2 | 114 |
