siRNA Details
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170468.1 | NP_001163939.1 | 5 | 708 | ||
XM_006251958.2 | XP_006252020.1 | 5 | 677 | |||
XM_006251959.2 | XP_006252021.1 | 5 | 677 | |||
XM_006251960.2 | XP_006252022.1 | 4 | 557 | |||
XM_006251962.2 | XP_006252024.1 | 4 | 366 | |||
GenBank mRNA | FM031419.1 | - | 5 | 487 | ||
FM033220.1 | - | 4 | 369 |
