siRNA Details
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170468.1 | NP_001163939.1 | 15 | 3119 | ||
XM_006251958.2 | XP_006252020.1 | 15 | 3052 | |||
XM_006251959.2 | XP_006252021.1 | 15 | 3049 | |||
XM_006251960.2 | XP_006252022.1 | 14 | 2968 | |||
XM_006251962.2 | XP_006252024.1 | 14 | 2777 | |||
XM_006251963.1 | XP_006252025.1 | 8 | 1092 | |||
XM_006251964.1 | XP_006252026.1 | 8 | 874 | |||
GenBank mRNA | BC091320.1 | - | 6 | 612 | ||
CA512047.1 | - | 7 | 704 | |||
EV770592.1 | - | 2 | 174 |
