siRNA Details
Gene Symbol: Pspc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025672.1 | NP_001020843.1 | 2 | 594 | ||
XM_006252065.2 | XP_006252127.1 | 2 | 636 | |||
GenBank mRNA | BC099204.1 | - | 2 | 594 | 7384173 |

More Information
Other Species Targeted

Gene Ontology Categories:
