siRNA Details
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107269.1 | NP_001100739.1 | 10 | 1265 | ||
XM_006252177.2 | XP_006252239.1 | 11 | 1301 | |||
XM_008770751.1 | XP_008768973.1 | 12 | 3178 | |||
XM_008770752.1 | XP_008768974.1 | 13 | 2771 |
