siRNA Details
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083337.1 | NP_001076806.1 | 3 | 202 | ||
XM_006252181.2 | XP_006252243.1 | 3 | 1105 | |||
GenBank mRNA | BC158553.1 | - | 3 | 301 | 9088212 | |
EF446165.1 | - | 3 | 202 | |||
FQ232228.1 | - | 2 | 90 |
