siRNA Details
Gene Symbol: Hmbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252219.2 | XP_006252281.1 | 3 | 667 | ||
XM_006252220.2 | XP_006252282.1 | 3 | 667 | |||
XM_006252221.2 | XP_006252283.1 | 3 | 666 | |||
XM_008770802.1 | XP_008769024.1 | 3 | 594 | |||
XM_008770803.1 | XP_008769025.1 | 3 | 667 | |||
XM_008770804.1 | XP_008769026.1 | 3 | 667 | |||
XM_008770805.1 | XP_008769027.1 | 3 | 667 |
