siRNA Details
Gene Symbol: Hmbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252219.2 | XP_006252281.1 | 10 | 1489 | ||
XM_006252220.2 | XP_006252282.1 | 10 | 1486 | |||
XM_008770802.1 | XP_008769024.1 | 10 | 1416 | |||
XM_008770803.1 | XP_008769025.1 | 10 | 1483 |
