siRNA Details
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013132.2 | NP_001013150.1 | 4 | 423 | ||
XM_006252201.2 | XP_006252263.1 | 5 | 1196 | |||
XM_006252202.2 | XP_006252264.1 | 4 | 1076 | |||
GenBank mRNA | BC062801.1 | - | 4 | 426 | 6889991 | |
FQ213407.1 | - | 4 | 357 |
