siRNA Details
Gene Symbol: Piwil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107276.1 | NP_001100746.1 | 16, 17 | 2177 | ||
XM_006252271.2 | XP_006252333.2 | 16, 17 | 2123 | |||
XM_008770828.1 | XP_008769050.1 | 15, 16 | 2013 |
