siRNA Details
Gene Symbol: Pcdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107279.1 | NP_001100749.1 | 2 | 2749 | ||
XM_006252377.2 | XP_006252439.1 | 1 | 2565 | |||
XM_006252378.2 | XP_006252440.1 | 1 | 2565 | |||
XM_006252379.1 | XP_006252441.1 | 1 | 2609 |
