siRNA Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012043.1 | NP_001012043.1 | 12 | 2218 | ||
GenBank mRNA | BC081929.1 | - | 12 | 2218 | 7113366 | |
FQ226413.1 | - | 2 | 848 | |||
FQ232347.1 | - | 2 | 848 |
