siRNA Details
Gene Symbol: Lcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012044.1 | NP_001012044.1 | 14 | 1620 | ||
XM_006252318.2 | XP_006252380.1 | 14 | 1795 | |||
XM_006252319.2 | XP_006252381.1 | 16 | 1994 | |||
XM_006252320.2 | XP_006252382.1 | 16 | 2042 | |||
XM_006252322.2 | XP_006252384.1 | 14 | 3201 | |||
GenBank mRNA | BC083855.1 | - | 14 | 1620 | 7125327 |
