siRNA Details
Gene Symbol: Lcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012044.1 | NP_001012044.1 | 7 | 741 | ||
XM_006252318.2 | XP_006252380.1 | 7 | 916 | |||
XM_006252319.2 | XP_006252381.1 | 9 | 1115 | |||
XM_006252320.2 | XP_006252382.1 | 9 | 1163 | |||
XM_006252322.2 | XP_006252384.1 | 7 | 2322 | |||
GenBank mRNA | BC083855.1 | - | 7 | 741 | 7125327 |
