siRNA Details
Gene Symbol: Rbm26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277160.1 | NP_001264089.1 | 18 | 2996 | ||
XM_006252422.1 | XP_006252484.1 | 19 | 2729 | |||
XM_006252424.1 | XP_006252486.1 | 19 | 2699 | |||
XM_006252425.1 | XP_006252487.1 | 19 | 2729 | |||
XM_006252426.1 | XP_006252488.1 | 18 | 2996 | |||
XM_006252427.1 | XP_006252489.1 | 18 | 2627 | |||
XM_008770872.1 | XP_008769094.1 | 19 | 2729 | |||
GenBank mRNA | BC082039.1 | - | 12 | 1727 |
