siRNA Details
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006994.1 | NP_001006995.1 | 6 | 815 | ||
XM_006252452.2 | XP_006252514.1 | 6 | 911 | |||
XM_006252455.2 | XP_006252517.1 | 4 | 681 | |||
GenBank mRNA | BC083689.1 | - | 6 | 815 | 7189368 |
