siRNA Details
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252478.2 | XP_006252540.1 | 10 | 1073 | ||
XM_224534.8 | XP_224534.7 | 11 | 1319 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252478.2 | XP_006252540.1 | 10 | 1073 | ||
XM_224534.8 | XP_224534.7 | 11 | 1319 |