siRNA Details
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013134.2 | NP_001013152.2 | 13 | 2200 | ||
XM_008770996.1 | XP_008769218.1 | 13 | 2304 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013134.2 | NP_001013152.2 | 13 | 2200 | ||
XM_008770996.1 | XP_008769218.1 | 13 | 2304 |