siRNA Details
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030037.1 | NP_001025208.1 | 7 | 898 | ||
XM_006252608.2 | XP_006252670.2 | 8 | 1131 | |||
XM_008770997.1 | XP_008769219.1 | 7 | 1702 | |||
GenBank mRNA | BC099770.1 | - | 7 | 898 | 7932056 |
