siRNA Details
Gene Symbol: Lrrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013999.1 | NP_001014021.1 | 3 | 324 | ||
XM_008771097.1 | XP_008769319.1 | 3 | 700 | |||
GenBank mRNA | BC081908.1 | - | 3 | 324 | 7111390 |

More Information
Other Species Targeted

