siRNA Details
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107308.1 | NP_001100778.1 | 4, 5 | 366 | ||
XM_006252903.2 | XP_006252965.1 | 4, 5 | 816 | |||
XM_008771109.1 | XP_008769331.1 | 4, 5 | 816 |
