siRNA Details
Gene Symbol: Cnot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107313.1 | NP_001100783.1 | 5 | 859 | ||
XM_008771290.1 | XP_008769512.1 | 5 | 1032 | |||
XM_008771291.1 | XP_008769513.1 | 5 | 1002 | |||
XM_008771292.1 | XP_008769514.1 | 5 | 902 | |||
GenBank mRNA | BC167766.1 | - | 5 | 818 | 7455944 |
