siRNA Details
Gene Symbol: LOC103693999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771409.1 | XP_008769631.1 | 12 | 2216 | ||
XM_008771410.1 | XP_008769632.1 | 12 | 2213 | |||
XM_008771411.1 | XP_008769633.1 | 11 | 2119 | |||
XM_008771412.1 | XP_008769634.1 | 11 | 2116 |
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135857.1 | NP_001129329.1 | 2233 | |||
XM_008771391.1 | XP_008769613.1 | 1901 | ||||
XM_008771392.1 | XP_008769614.1 | 1898 | ||||
XM_008771393.1 | XP_008769615.1 | 1804 | ||||
XM_008771394.1 | XP_008769616.1 | 1801 |
