siRNA Details
Gene Symbol: Myom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169141.1 | NP_001162612.1 | 15 | 1814 | ||
XM_006253389.2 | XP_006253451.1 | 15 | 1915 | |||
XM_008771360.1 | XP_008769582.1 | 15 | 1732 |
