siRNA Details
Gene Symbol: Abhd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271072.1 | NP_001258001.1 | 2 | 648 | ||
XM_006253478.2 | XP_006253540.1 | 2 | 437 | |||
XM_006253479.2 | XP_006253541.1 | 2 | 521 | |||
XM_008771395.1 | XP_008769617.1 | 3 | 593 | |||
XM_008771396.1 | XP_008769618.1 | 2 | 468 | |||
GenBank mRNA | FQ212188.1 | - | 2 | 632 | ||
FQ221944.1 | - | 2 | 617 |
