siRNA Details
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107337.1 | NP_001100807.1 | 11 | 4539 | ||
XM_006253620.2 | XP_006253682.1 | 11 | 4788 | |||
XM_006253621.2 | XP_006253683.1 | 11 | 4890 | |||
XM_006253622.2 | XP_006253684.1 | 11 | 4618 | |||
XM_006253623.2 | XP_006253685.1 | 11 | 4777 | |||
XM_006253624.2 | XP_006253686.1 | 11 | 4714 | |||
XM_006253625.2 | XP_006253687.1 | 11 | 4893 | |||
XM_006253627.2 | XP_006253689.1 | 11 | 4787 | |||
XM_008771491.1 | XP_008769713.1 | 11 | 4843 | |||
XM_008771492.1 | XP_008769714.1 | 10 | 3754 | |||
XM_008771493.1 | XP_008769715.1 | 10 | 4044 | |||
XM_008771494.1 | XP_008769716.1 | 11 | 4787 |
