siRNA Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008556.1 | NP_001008556.1 | 13 | 1262 | ||
XM_008771534.1 | XP_008769756.1 | 12 | 1977 | |||
XM_008771535.1 | XP_008769757.1 | 13 | 1361 | |||
GenBank mRNA | AY823319.1 | - | 13 | 1262 |
