siRNA Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008556.1 | NP_001008556.1 | 10 | 948 | ||
XM_008771534.1 | XP_008769756.1 | 9 | 1663 | |||
XM_008771535.1 | XP_008769757.1 | 10 | 1047 | |||
GenBank mRNA | AY823319.1 | - | 10 | 948 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001276399, NM_199056, BC065157, AK160092, AK167467, and others.
Gene Ontology Categories:
