siRNA Details
Gene Symbol: Wnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191556.1 | NP_001178485.1 | 4 | 1210 | ||
XM_006253715.2 | XP_006253777.1 | 4 | 1134 | |||
XM_006253720.2 | XP_006253782.1 | 4 | 1134 | |||
XM_006253724.1 | XP_006253786.1 | 4 | 1134 | |||
XM_006253725.2 | XP_006253787.1 | 4 | 1134 | |||
XM_006253727.1 | XP_006253789.1 | 4 | 1134 | |||
XM_008771536.1 | XP_008769758.1 | 4 | 1134 | |||
XM_008771537.1 | XP_008769759.1 | 4 | 1134 | |||
XM_008771538.1 | XP_008769760.1 | 4 | 8450 |
