siRNA Details
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012052.1 | NP_001012052.1 | 6 | 2074 | ||
XM_006253808.2 | XP_006253870.1 | 6 | 2061 | |||
XM_006253809.2 | XP_006253871.1 | 5 | 1752 | |||
GenBank mRNA | BC081934.1 | - | 6 | 2074 | 7114574 |
