siRNA Details
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012052.1 | NP_001012052.1 | 9 | 2420 | ||
XM_006253808.2 | XP_006253870.1 | 9 | 2407 | |||
XM_006253809.2 | XP_006253871.1 | 8 | 2098 | |||
XM_006253810.2 | XP_006253872.1 | 8 | 2290 | |||
XM_006253811.2 | XP_006253873.1 | 8 | 2374 | |||
GenBank mRNA | BC081934.1 | - | 9 | 2420 | 7114574 | |
FQ211932.1 | - | 8 | 2378 |
