siRNA Details
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107353.1 | NP_001100823.1 | 3 | 503 | ||
XM_006253966.2 | XP_006254028.1 | 3 | 580 | |||
XM_006253967.2 | XP_006254029.1 | 3 | 327 | |||
XM_006253968.2 | XP_006254030.1 | 3 | 596 | |||
XM_006253969.2 | XP_006254031.1 | 3 | 580 | |||
XM_008771654.1 | XP_008769876.1 | 3 | 1764 | |||
XM_008771655.1 | XP_008769877.1 | 3 | 1511 | |||
XM_008771656.1 | XP_008769878.1 | 3 | 1764 |
