siRNA Details
Gene Symbol: Mtpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107359.1 | NP_001100829.1 | 5 | 986 | ||
XM_006254071.2 | XP_006254133.1 | 5 | 1089 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mtpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107359.1 | NP_001100829.1 | 5 | 986 | ||
XM_006254071.2 | XP_006254133.1 | 5 | 1089 |