siRNA Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039023.1 | NP_001034112.1 | 7 | 928 | ||
XM_006254206.2 | XP_006254268.1 | 9 | 1146 | |||
GenBank mRNA | BC100154.1 | - | 7 | 928 | 7460511 |
