siRNA Details
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107367.2 | NP_001100837.1 | 6 | 913 | ||
XM_008771868.1 | XP_008770090.1 | 6 | 1322 | |||
XM_008771869.1 | XP_008770091.1 | 6 | 1320 | |||
XM_008771870.1 | XP_008770092.1 | 4 | 1060 | |||
GenBank mRNA | BC162024.1 | - | 6 | 913 | 9099401 |
