siRNA Details
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271281.1 | NP_001258210.1 | 13, 14 | 1119 | ||
XM_008772138.1 | XP_008770360.1 | 12, 13 | 1372 | |||
GenBank mRNA | CK364772.1 | - | 5, 6 | 344 |
