siRNA Details
Gene Symbol: Zfp532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107382.1 | NP_001100852.1 | 9 | 3544 | ||
XM_006254865.2 | XP_006254927.1 | 9 | 5834 | |||
XM_006254866.2 | XP_006254928.1 | 9 | 5273 | |||
XM_006254868.2 | XP_006254930.1 | 9 | 3861 | |||
XM_006254869.2 | XP_006254931.1 | 9 | 3706 | |||
XM_006254870.2 | XP_006254932.1 | 9 | 5238 | |||
XM_006254871.2 | XP_006254933.1 | 8 | 3419 | |||
XM_006254872.2 | XP_006254934.1 | 9 | 4618 | |||
XM_006254873.2 | XP_006254935.1 | 8 | 5724 | |||
XM_006254875.2 | XP_006254937.1 | 5 | 3938 | |||
XM_006254876.2 | XP_006254938.1 | 4 | 3656 | |||
XM_008772142.1 | XP_008770364.1 | 8 | 4460 |
