siRNA Details
Gene Symbol: Zfp532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107382.1 | NP_001100852.1 | 3 | 793 | ||
XM_006254865.2 | XP_006254927.1 | 3 | 3083 | |||
XM_006254866.2 | XP_006254928.1 | 3 | 2522 | |||
XM_006254868.2 | XP_006254930.1 | 3 | 1110 | |||
XM_006254869.2 | XP_006254931.1 | 3 | 955 | |||
XM_006254870.2 | XP_006254932.1 | 3 | 2487 | |||
XM_006254871.2 | XP_006254933.1 | 2 | 668 | |||
XM_006254872.2 | XP_006254934.1 | 3 | 1867 | |||
XM_006254873.2 | XP_006254935.1 | 2 | 2973 | |||
XM_006254875.2 | XP_006254937.1 | 2 | 1709 | |||
XM_006254876.2 | XP_006254938.1 | 2 | 1709 | |||
XM_008772142.1 | XP_008770364.1 | 2 | 1709 |
