siRNA Details
Gene Symbol: Ablim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191698.1 | NP_001178627.1 | 9, 10 | 1263 | ||
XM_008772143.1 | XP_008770365.1 | 9, 10 | 1332 | |||
XM_008772144.1 | XP_008770366.1 | 9, 10 | 1332 |
