siRNA Details
Gene Symbol: Ablim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191698.1 | NP_001178627.1 | 12 | 1514 | ||
XM_008772143.1 | XP_008770365.1 | 12 | 1583 | |||
XM_008772144.1 | XP_008770366.1 | 12 | 1583 | |||
XM_008772145.1 | XP_008770367.1 | 10 | 1397 | |||
XM_008772146.1 | XP_008770368.1 | 10 | 1397 | |||
GenBank mRNA | FQ217083.1 | - | 6 | 492 |
