siRNA Details
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007689.1 | NP_001007690.1 | 2 | 434 | ||
XM_006255106.2 | XP_006255168.1 | 2 | 521 | |||
XM_006255107.2 | XP_006255169.1 | 2 | 233 | |||
XM_006255108.2 | XP_006255170.1 | 2 | 236 | |||
GenBank mRNA | BC083753.1 | - | 2 | 434 | 7192604 | |
FQ211546.1 | - | 2 | 218 | |||
FQ213033.1 | - | 2 | 265 | |||
FQ230885.1 | - | 2 | 446 |
