siRNA Details
Gene Symbol: Rpgrip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107414.1 | NP_001100884.1 | 10 | 1193 | ||
XM_008772313.1 | XP_008770535.1 | 9 | 1621 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rpgrip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107414.1 | NP_001100884.1 | 10 | 1193 | ||
XM_008772313.1 | XP_008770535.1 | 9 | 1621 |