siRNA Details
Gene Symbol: Sall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107415.2 | NP_001100885.2 | 2 | 2757 | ||
XM_006255259.2 | XP_006255321.1 | 3 | 3291 | |||
XM_006255260.2 | XP_006255322.1 | 3 | 2882 | |||
XM_008772401.1 | XP_008770623.1 | 2 | 5763 |
