siRNA Details
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003653.1 | NP_001003653.1 | 3 | 1048 | ||
NM_001003654.1 | NP_001003654.1 | 3 | 1080 | |||
XM_006255490.2 | XP_006255552.1 | 3 | 1099 | |||
GenBank mRNA | AY682586.1 | - | 3 | 1080 | ||
AY682587.1 | - | 3 | 1048 | |||
DQ011518.1 | - | 3 | 1016 |
