siRNA Details
Gene Symbol: Gfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107421.1 | NP_001100891.1 | 3 | 904 | ||
XM_008772497.1 | XP_008770719.1 | 3 | 1958 | |||
XM_008772498.1 | XP_008770720.1 | 3 | 1498 | |||
GenBank mRNA | BC161901.1 | - | 3 | 886 | 8367133 |
